Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Glu(Met-OH)-OH TFA | 99.84% | C12H19F3N2O7S | 1 ML
SDP

Catalog No. 5000353879
Encompass_Preferred

H-Glu(Met-OH)-OH TFA (γ-Glu-Met TFA) could induce oxidation of hydroxyl radical. It is intended for research use only and not for sale to patients.

  • Molecular weight: 392.35
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: H-Glu(Met-OH)-OH
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Sealed storage, away from moisture

Catalog No. 50-003-53879 Supplier Medchemexpress LLC Supplier No. HY145779A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Glu(Met-OH)-OH TFA (γ-Glu-Met TFA) could induce oxidation of hydroxyl radical. It is intended for research use only and not for sale to patients.

  • Molecular weight: 392.35
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: H-Glu(Met-OH)-OH
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Sealed storage, away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.