Learn More
Apexbio Technology LLC H-Glu(OEt)-OEt·HCl 1118-89-4 25g
SDP
Catalog No. 5000093164
Encompass_Preferred
H-Glu OEt -OEt-HCl (CAS 1118-89-4) is an ethyl ester derivative of glutamate It is designed to serve as a glutamate precursor facilitating controlled intracellular glutamate availability and modulating glutamatergic signaling pathways H-Glu OEt -OEt-HCl exerts its biological activity primarily through enhanced membrane permeability followed by intracellular esterase-mediated conversion to free glutamate Based on these pharmacological properties H-Glu OEt -OEt-HCl holds research potential in neuroscience and biochemical pharmacology particularly for studying glutamate-dependent neuronal signaling synaptic transmission and mechanisms underlying neurodegenerative disorders
Catalog No. 50-000-93164 Supplier Apexbio Technology LLC Supplier No. A717025000
H-Glu OEt -OEt-HCl (CAS 1118-89-4) is an ethyl ester derivative of glutamate It is designed to serve as a glutamate precursor facilitating controlled intracellular glutamate availability and modulating glutamatergic signaling pathways H-Glu OEt -OEt-HCl exerts its biological activity primarily through enhanced membrane permeability followed by intracellular esterase-mediated conversion to free glutamate Based on these pharmacological properties H-Glu OEt -OEt-HCl holds research potential in neuroscience and biochemical pharmacology particularly for studying glutamate-dependent neuronal signaling synaptic transmission and mechanisms underlying neurodegenerative disorders