Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Glu(OtBu)-NH2 hydrochloride | 108607-02-9 | 98.0% | 238.71 g·mol-1 | C9H19ClN2O3 | 25 G
SDP

Catalog No. 5000209622
Encompass_Preferred

H-Glu(OtBu)-NH2 hydrochloride is a tert-butyl-protected glutamic acid amide provided as the hydrochloride salt for research use. It is supplied as a solid reagent commonly used in peptide synthesis and related organic chemistry applications where a protected glutamic acid amide is required.

  • Tert-butyl protection on the glutamic acid side chain
  • Suitable for peptide synthesis and organic synthesis workflows
  • High purity (98.0%) for reliable reactions
  • Available in a 25 G pack for multi-gram scale work
  • Identified by CAS number 108607-02-9
  • Molecular weight 238.71 g·mol-1; formula C9H19ClN2O3
  • Room temperature storage recommended for long-term stability; in solvent store at -20°C to -80°C

Catalog No. 50-002-09622 Supplier Medchemexpress LLC Supplier No. 25GHYW088929
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Glu(OtBu)-NH2 hydrochloride is a tert-butyl-protected glutamic acid amide provided as the hydrochloride salt for research use. It is supplied as a solid reagent commonly used in peptide synthesis and related organic chemistry applications where a protected glutamic acid amide is required.

  • Tert-butyl protection on the glutamic acid side chain
  • Suitable for peptide synthesis and organic synthesis workflows
  • High purity (98.0%) for reliable reactions
  • Available in a 25 G pack for multi-gram scale work
  • Identified by CAS number 108607-02-9
  • Molecular weight 238.71 g·mol-1; formula C9H19ClN2O3
  • Room temperature storage recommended for long-term stability; in solvent store at -20°C to -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.