Learn More
Medchemexpress LLC H-Glu(OtBu)-OtBu hydrochloride | 32677-01-3 | 98.0% | 295.80 | 500 G
SDP
Catalog No. 5000401201
Encompass_Preferred
H-Glu(OtBu)-OtBu hydrochloride is a key intermediate utilized in the synthesis of prostate-specific membrane antigen (PSMA) targeting probes. This compound has the ability to reduce nonspecific background binding through negatively charged linkers, thereby improving tumor/background contrast. It is also applicable in prostate cancer PET/SPECT imaging studies.
  • Key intermediate in PSMA targeting probe synthesis.
  • Reduces nonspecific background binding.
  • Improves tumor/background contrast.
  • Applicable in prostate cancer PET/SPECT imaging studies.
  • Can be used to synthesize PSMA probe 5 for tumor imaging.
Catalog No. 50-004-01201 Supplier Medchemexpress LLC Supplier No. HY20167A500G
H-Glu(OtBu)-OtBu hydrochloride is a key intermediate utilized in the synthesis of prostate-specific membrane antigen (PSMA) targeting probes. This compound has the ability to reduce nonspecific background binding through negatively charged linkers, thereby improving tumor/background contrast. It is also applicable in prostate cancer PET/SPECT imaging studies.
  • Key intermediate in PSMA targeting probe synthesis.
  • Reduces nonspecific background binding.
  • Improves tumor/background contrast.
  • Applicable in prostate cancer PET/SPECT imaging studies.
  • Can be used to synthesize PSMA probe 5 for tumor imaging.