Learn More
Medchemexpress LLC H-Glu-Pro-Leu-Gln-Leu-Lys-Met-OH acetate | 918.11 | 50 MG
SDP
Catalog No. 5000397275
Encompass_Preferred
H-Glu-Pro-Leu-Gln-Leu-Lys-Met-OH acetate (EM7) is a linear mesenchymal stem cell (MSC) specific peptide. It can be used to prepare Gd-DOTA-peptide complexes.
  • Linear mesenchymal stem cell (MSC) specific peptide
  • Used to prepare Gd-DOTA-peptide complexes
  • Store sealed, away from moisture
  • Powder storage at -80°C for 2 years, or -20°C for 1 year
  • In-solvent storage at -80°C for 6 months, or -20°C for 1 month
  • Soluble in H2O at ≥ 50 mg/mL (54.46 mM)
  • Related classifications include peptides and peptide derivatives
  • Related classifications include peptides for drug delivery and targeting peptides
Catalog No. 50-003-97275 Supplier Medchemexpress LLC Supplier No. HYP3413A50MG
H-Glu-Pro-Leu-Gln-Leu-Lys-Met-OH acetate (EM7) is a linear mesenchymal stem cell (MSC) specific peptide. It can be used to prepare Gd-DOTA-peptide complexes.
  • Linear mesenchymal stem cell (MSC) specific peptide
  • Used to prepare Gd-DOTA-peptide complexes
  • Store sealed, away from moisture
  • Powder storage at -80°C for 2 years, or -20°C for 1 year
  • In-solvent storage at -80°C for 6 months, or -20°C for 1 month
  • Soluble in H2O at ≥ 50 mg/mL (54.46 mM)
  • Related classifications include peptides and peptide derivatives
  • Related classifications include peptides for drug delivery and targeting peptides