Learn More
Bachem H-Gly-2-chlorotrityl resin (200-400 mesh, 0.50-0.90 mmol/g)
SDP
Catalog No. 50258911
Encompass_Preferred
H-Gly-2-chlorotrityl resin (200-400 mesh, 0.50-0.90 mmol/g), 5g.
Catalog No. 50-258-911 Supplier Bachem Supplier No. 4026823.0005
H-Gly-2-chlorotrityl resin (200-400 mesh, 0.50-0.90 mmol/g), 5g.