Learn More
Bachem H-Gly-Arg-Gly-Asp-Asn-Pro-OH
SDP
Supplier:  Bachem 4030603.0025
Encompass_Preferred
H-Gly-Arg-Gly-Asp-Asn-Pro-OH, 114681-65-1, C23H38N10O10, Mr 614.62, 25mg. GRGDNP, G-R-G-D-N-P. Potent inhibitor of cell attachment to fibronectin. GRGDNP promotes cell adhesion preferentially via the a5b1 integrin.
Catalog No. 50-382-711