Learn More
Bachem H-Gly-Arg-Gly-Asp-Thr-Pro-OH
SDP
Supplier:  Bachem 4012897.0005
Encompass_Preferred
H-Gly-Arg-Gly-Asp-Thr-Pro-OH, 108682-58-2, C23H39N9O10, Mr 601.62, 5mg. GRGDTP, G-R-G-D-T-P. GRGDTP prevents cell attachment of fibronectin.
Catalog No. 50-383-376