Learn More
Bachem H-Gly-Arg-Gly-Glu-Ser-Pro-OH
SDP
Supplier:  Bachem 4030609.0005
Encompass_Preferred
H-Gly-Arg-Gly-Glu-Ser-Pro-OH Trifluoroacetate, 99896-88-5, C23H39N9O10, Mr 601.62, 5mg. GRGESP, G-R-G-E-S-P. Inactive fibronectin fragment analog, used as control for tests with GRGDSP.
Catalog No. 50-259-709