Learn More
Bachem H-Gly-Asp-Asp-Asp-Asp-Lys-bNA
SDP
Supplier:  Bachem 4001598.0025
Encompass_Preferred
H-Gly-Asp-Asp-Asp-Asp-Lys-bNA Ammonium, 70023-02-8, C34H44N8O14, Mr 788.77, 25mg. GDDDDK-bNA, specific substrate for the determination of enteropeptidase (enterokinase), also used in histochemistry.
Catalog No. 50-260-369