Learn More
Medchemexpress LLC H-Gly-Leu-Phe-OH | 103213-38-3 | 99.4% | 335.40 | 5 MG
SDP
Catalog No. 5000399736
Encompass_Preferred
H-Gly-Leu-Phe-OH (GLF) is an immunostimulatory tripeptide derived from α-lactalbumin. It inhibits anticancer agent Etoposide-induced alopecia, epidermal thickening, and thinning of the adipocyte layer. This product is intended for research use only.
  • Immunostimulatory tripeptide
  • Derived from α-lactalbumin
  • Inhibits etoposide-induced alopecia, epidermal thickening, and thinning of the adipocyte layer
  • For research use only
  • EC50: 0.28 mM for activation of human PEPT1 expressed in MDCK cells
  • IC50: 0.98 mM for binding affinity to human PEPT1 assessed as inhibition of [14C]Gly-Sar uptake in MDCK cells
Catalog No. 50-003-99736 Supplier Medchemexpress LLC Supplier No. HYP42865MG
H-Gly-Leu-Phe-OH (GLF) is an immunostimulatory tripeptide derived from α-lactalbumin. It inhibits anticancer agent Etoposide-induced alopecia, epidermal thickening, and thinning of the adipocyte layer. This product is intended for research use only.
  • Immunostimulatory tripeptide
  • Derived from α-lactalbumin
  • Inhibits etoposide-induced alopecia, epidermal thickening, and thinning of the adipocyte layer
  • For research use only
  • EC50: 0.28 mM for activation of human PEPT1 expressed in MDCK cells
  • IC50: 0.98 mM for binding affinity to human PEPT1 assessed as inhibition of [14C]Gly-Sar uptake in MDCK cells