Learn More
Bachem H-Gly-Pro-Ala-OH
SDP
Supplier:  Bachem 4002533.0001
Encompass_Preferred
H-Gly-Pro-Ala-OH, 837-83-2, C10H17N3O4, Mr 243.26, 1g. Cleavage product of the collagenase substrate Z-GPGGPA. The collagen tripeptide Inhibits dipeptidylpeptidase IV (DPP IV).
Catalog No. 50-259-761