Learn More
Bachem H-Gly-Pro-AMC · HBr
SDP
Catalog No. 50260289
Encompass_Preferred
H-Gly-Pro-AMC · HBr, 115035-46-6, C17H19N3O4 · HBr, Mr 410.27, 25mg. Highly sensitive substrate for dipeptidyl aminopeptidase IV (DPP IV, X-prolyldipeptidyl aminopeptidase).
Catalog No. 50-260-289 Supplier Bachem Supplier No. 4002520.0025
Add to Cart
Add to Cart
H-Gly-Pro-AMC · HBr, 115035-46-6, C17H19N3O4 · HBr, Mr 410.27, 25mg. Highly sensitive substrate for dipeptidyl aminopeptidase IV (DPP IV, X-prolyldipeptidyl aminopeptidase).