Learn More
Bachem H-Gly-Pro-Arg-Pro-OH
SDP
Catalog No. 50382652
Encompass_Preferred
H-Gly-Pro-Arg-Pro-OH Acetate, 67869-62-9, C18H31N7O5, Mr 425.49, 25mg. Fibrinolysis Inhibiting Factor. GPRP (H-2935) inhibits fibrinogen/thrombin clotting, though it is less active than its C-terminal amide GPRP-NH2.
Catalog No. 50-382-652 Supplier Bachem Supplier No. 4004545.0025
Add to Cart
Add to Cart
H-Gly-Pro-Arg-Pro-OH Acetate, 67869-62-9, C18H31N7O5, Mr 425.49, 25mg. Fibrinolysis Inhibiting Factor. GPRP (H-2935) inhibits fibrinogen/thrombin clotting, though it is less active than its C-terminal amide GPRP-NH2.