Learn More
Carbosynth LLC. H-Gly-Tyr-Pro-Gly-Gln-Val-NH2, 1mg, MW: (M.W. 618.70), Molecular formula: C28H42N8O8
SDP
Supplier:  Carbosynth LLC. PAR3673PI1MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: H-Gly-Tyr-Pro-Gly-Gln-Val-Amide, GYPGQV-Amide, H-GYPGQV-NH2, Application: Tethered Ligand (Human) / Protease-Activated Receptor 4 (PAR4) Agonist, Selective PAR4 Agonist, Salt Form:, Acetate Salt, Storage Temperature: -20C, References: W. Xu, H. Andersen, T.E. Whitmore, S.R. Presnell, D.P. Yee, A. Ching, T. Gilbert, E.W. Davie, and D.C. Foster, Proc. Nat 'l. Acad. Sci. U.S.A., 95, 6642 (1998)., M.D. Hollenberg, M. Saifeddine, B. Al-Ani, and Y. Gui, Can. J. Physiol. Pharmacol., 77, 458 (1999)., N. Vergnolle, Br. J. Pharmacol., 141, 1264 (2004). (Review)
Catalog No. 50-168-6180