Learn More
Medchemexpress LLC HY-P1226 1mg Medchemexpress, HAEGTFTSD CAS:926018-45-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP12261MG
Encompass_Preferred
Medchemexpress, HY-P1226 1mg HAEGTFTSD CAS:926018-45-3 HAEGTFTSD is the first N-terminal 1-9 residues of GLP-1 peptide. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4167