Learn More
Medchemexpress LLC HY-P1224 1mg Medchemexpress, HAEGTFTSDVS CAS:864915-61-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP12241MG
Encompass_Preferred
Medchemexpress, HY-P1224 1mg HAEGTFTSDVS CAS:864915-61-7 HAEGTFTSDVS is the first N-terminal 1-11 residues of GLP-1 peptide. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4165