Learn More
Medchemexpress LLC HY-P1445 1mg Medchemexpress, HAEGTFTSDVSSYLE CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP14451MG
Encompass_Preferred
Medchemexpress, HY-P1445 1mg HAEGTFTSDVSSYLE CAS: HAEGTFTSDVSSYLE is a polypeptide from patent CN 102920658 B. GLP-I analog contains the sequence. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4197