Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-HoArg-OH | 156-86-5 | 188.23 | 1 G
SDP

Catalog No. 5000446885
Encompass_Preferred

H-HoArg-OH is a homologue of arginine and a strong inhibitor of human bone and liver alkaline phosphatase, with virtually no effect on intestinal and placental alkaline phosphatases. Low levels of this endogenous amino acid have been linked to cardiovascular disease.

  • Formula: C7H16N4O2
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite, disease markers, endocrine diseases, nervous system disorder
  • Target: Alkaline phosphatase

Catalog No. 50-004-46885 Supplier Medchemexpress LLC Supplier No. HYW0083851G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-HoArg-OH is a homologue of arginine and a strong inhibitor of human bone and liver alkaline phosphatase, with virtually no effect on intestinal and placental alkaline phosphatases. Low levels of this endogenous amino acid have been linked to cardiovascular disease.

  • Formula: C7H16N4O2
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Endogenous metabolite, disease markers, endocrine diseases, nervous system disorder
  • Target: Alkaline phosphatase

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.