Learn More
Medchemexpress LLC H-HoArg-OH | 156-86-5 | 188.23 | 10 G
SDP
Catalog No. 5000447314
Encompass_Preferred
H-HoArg-OH is a homologue arginine that acts as a strong inhibitor of human bone and liver alkaline phosphatase. Low levels of this endogenous amino acid have been linked to cardiovascular disease. This product is intended for research use only and is supplied as a white solid.
  • Strong inhibitor of human bone and liver alkaline phosphatase
  • Virtually no effect on intestinal and placental alkaline phosphatases
  • Supplied as a white solid
  • Linked to cardiovascular disease at low endogenous levels
Catalog No. 50-004-47314 Supplier Medchemexpress LLC Supplier No. HYW00838510G
H-HoArg-OH is a homologue arginine that acts as a strong inhibitor of human bone and liver alkaline phosphatase. Low levels of this endogenous amino acid have been linked to cardiovascular disease. This product is intended for research use only and is supplied as a white solid.
  • Strong inhibitor of human bone and liver alkaline phosphatase
  • Virtually no effect on intestinal and placental alkaline phosphatases
  • Supplied as a white solid
  • Linked to cardiovascular disease at low endogenous levels