Learn More
Apexbio Technology LLC H-HoArg-OH 156-86-5 25g
SDP
Catalog No. 5000093613
Encompass_Preferred
H-HoArg-OH (CAS 156-86-5) is a small molecule structurally analogous to arginine recognized for its inhibitory activity against human alkaline phosphatase (ALP) It exerts its effect by binding to the active site of ALP thereby impeding the enzyme s ability to hydrolyze phosphate ester substrates This interference modulates enzymatic processes and is valuable in investigating ALP s physiological functions and pathophysiological roles H-HoArg-OH serves as a research tool to explore ALP enzymology and related signaling pathways supporting studies focused on the development of therapeutics targeting ALP-associated disorders
Catalog No. 50-000-93613 Supplier Apexbio Technology LLC Supplier No. A754325000
H-HoArg-OH (CAS 156-86-5) is a small molecule structurally analogous to arginine recognized for its inhibitory activity against human alkaline phosphatase (ALP) It exerts its effect by binding to the active site of ALP thereby impeding the enzyme s ability to hydrolyze phosphate ester substrates This interference modulates enzymatic processes and is valuable in investigating ALP s physiological functions and pathophysiological roles H-HoArg-OH serves as a research tool to explore ALP enzymology and related signaling pathways supporting studies focused on the development of therapeutics targeting ALP-associated disorders