Learn More
Medchemexpress LLC HY-W012734 100mg Medchemexpress, H-HoPro-OH CAS:3105-95-1 Purity:>98%
SDP
Catalog No. 501934569
Encompass_Preferred
Medchemexpress, HY-W012734 100mg H-HoPro-OH CAS:3105-95-1 H-HoPro-OH is a breakdown product of lysine, accumulates in body fluids of infants with generalized genetic peroxisomal disorders, such as Zellweger syndrome, neonatal adrenoleukodystrophy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4569 Supplier Medchemexpress LLC Supplier No. HYW012734100MG
Add to Cart
Add to Cart
Medchemexpress, HY-W012734 100mg H-HoPro-OH CAS:3105-95-1 H-HoPro-OH is a breakdown product of lysine, accumulates in body fluids of infants with generalized genetic peroxisomal disorders, such as Zellweger syndrome, neonatal adrenoleukodystrophy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.