Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-hyp-ome hydrochloride | 40216-83-9 | 98.0% | 181.62 g/mol | C6H12ClNO3 | 500 G
SDP

Catalog No. 5000202787
Encompass_Preferred

H-Hyp-OMe hydrochloride is the hydrochloride salt of the methyl ester of trans-4-hydroxy-L-proline, used as a non-cleavable ADC linker, a PROTAC linker building block, and a synthetic intermediate in medicinal chemistry.

  • Hydrochloride salt form suitable for conjugation chemistry.
  • Molecular weight 181.62 g/mol; formula C6H12ClNO3.
  • Purity 98.0%.
  • Available in bulk 500 g pack.
  • SDS and COA PDFs available for handling and quality data.

Catalog No. 50-002-02787 Supplier Medchemexpress LLC Supplier No. HY76043500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Hyp-OMe hydrochloride is the hydrochloride salt of the methyl ester of trans-4-hydroxy-L-proline, used as a non-cleavable ADC linker, a PROTAC linker building block, and a synthetic intermediate in medicinal chemistry.

  • Hydrochloride salt form suitable for conjugation chemistry.
  • Molecular weight 181.62 g/mol; formula C6H12ClNO3.
  • Purity 98.0%.
  • Available in bulk 500 g pack.
  • SDS and COA PDFs available for handling and quality data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.