Learn More
Medchemexpress LLC H-Ile-Pro-Pro-OH hyd | 100MG
SDP
Catalog No. 5000228360
Encompass_Preferred
H-Ile-Pro-Pro-OH hydrochloride a milk-derived peptide[1] inhibits angiotensin-converting enzyme (ACE)[1] with an IC50 of 5 M[2] Antihypertensive tripeptides[1]
Catalog No. 50-002-28360 Supplier Medchemexpress LLC Supplier No. HY114424A100MG
H-Ile-Pro-Pro-OH hydrochloride a milk-derived peptide[1] inhibits angiotensin-converting enzyme (ACE)[1] with an IC50 of 5 M[2] Antihypertensive tripeptides[1]