Learn More
Gentarget h KLRK1 (CD314) Expression Lentivirus, Amount: 200ul, at 1 x 107 IFU/ml in DMEM medium with 10% FBS and 10x of polybrene (60 ug/ml).
Catalog No. NC3312608
Encompass
Lentivirus expresses human target: KLRK1 (CD314) gene (killer cell lectin like receptor K1), (Gene_Synonym: CD314; D12S2489E; KLR; NKG2-D; NKG2D), with coding sequence 100% matching to CDS region in NCBI ID: NM_007360. It contains a RFP–Blasticidin dual selection marker.
Catalog No. NC3312608 Supplier Gentarget Supplier No. LVP1377
Lentivirus expresses human target: KLRK1 (CD314) gene (killer cell lectin like receptor K1), (Gene_Synonym: CD314; D12S2489E; KLR; NKG2-D; NKG2D), with coding sequence 100% matching to CDS region in NCBI ID: NM_007360. It contains a RFP–Blasticidin dual selection marker.