Learn More
Medchemexpress LLC H-l-lys(4-n3-z)-oh (hydrochloride) | 2084913-49-3 | MFCD14706695 | 99.7% | 357.79 | C14H20ClN5O4 | 10 MG
SDP
Catalog No. 5000286503
Encompass_Preferred
H-L-Lys(4-N3-Z)-OH hydrochloride is an azide-bearing lysine derivative supplied as the hydrochloride salt for use as a bioorthogonal click-chemistry reagent and probe. The azide moiety enables selective ligation via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC), facilitating efficient bioconjugation in synthetic and biological workflows.
  • Azide functional group enables CuAAC and SPAAC.
  • Designed for selective bioconjugation and probe attachment.
  • Suitable for probe synthesis and linker development for conjugates.
  • Supplied as hydrochloride salt for improved handling and stability.
  • High reported purity (≈99.7%) for analytical and synthetic use.
Catalog No. 50-002-86503 Supplier Medchemexpress LLC Supplier No. HY15176110MG
H-L-Lys(4-N3-Z)-OH hydrochloride is an azide-bearing lysine derivative supplied as the hydrochloride salt for use as a bioorthogonal click-chemistry reagent and probe. The azide moiety enables selective ligation via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted alkyne-azide cycloaddition (SPAAC), facilitating efficient bioconjugation in synthetic and biological workflows.
  • Azide functional group enables CuAAC and SPAAC.
  • Designed for selective bioconjugation and probe attachment.
  • Suitable for probe synthesis and linker development for conjugates.
  • Supplied as hydrochloride salt for improved handling and stability.
  • High reported purity (≈99.7%) for analytical and synthetic use.