Learn More
Medchemexpress LLC HY-D0853 5mg Medchemexpress, H-L-Photo-lysine CAS:1253643-88-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD08535MG
Encompass_Preferred
Medchemexpress, HY-D0853 5mg H-L-Photo-lysine CAS:1253643-88-7 H-L-Photo-lysine is a diazirine-containing lysine amino acid and is a photo-cross-linker. H-L-Photo-lysine can site-selective incorporated into proteins and is used to crosslink protein-protein interactions in vitro and in living cells. H-L-Photo-lysine acts as a UV light-activated photo-crosslinking probe. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2748