Learn More
Medchemexpress LLC HY-D0853A 5mg Medchemexpress, H-L-Photo-lysine (hydrochloride) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD0853A5MG
Encompass_Preferred
Medchemexpress, HY-D0853A 5mg H-L-Photo-lysine (hydrochloride) CAS:0 H-L-Photo-lysine hydrochloride is a diazirine-containing lysine amino acid and is a photo-cross-linker. H-L-Photo-lysine hydrochloride can site-selective incorporated into proteins and is used to crosslink protein-protein interactions in vitro and in living cells. H-L-Photo-lysine hydrochloride acts as a UV light-activated photo-crosslinking probe[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1271