Learn More
Medchemexpress LLC H-L-Tyr(2-azidoethyl)-OH | 1570523-47-5 | C11H14N4O3 | 50 MG
SDP
Catalog No. 5000372175
Encompass_Preferred
H-L-Tyr(2-azidoethyl)-OH is an unnatural Tyrosine derivative and a click chemistry reagent containing an azide group. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups, and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups. This product is for research use only. Please store the product under the recommended conditions in the Certificate of Analysis; shipping is at room temperature in continental US and may vary elsewhere.
  • Unnatural Tyrosine derivative
  • Click chemistry reagent containing an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Undergoes ring strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-72175 Supplier Medchemexpress LLC Supplier No. HY15169350MG
H-L-Tyr(2-azidoethyl)-OH is an unnatural Tyrosine derivative and a click chemistry reagent containing an azide group. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups, and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups. This product is for research use only. Please store the product under the recommended conditions in the Certificate of Analysis; shipping is at room temperature in continental US and may vary elsewhere.
  • Unnatural Tyrosine derivative
  • Click chemistry reagent containing an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Undergoes ring strain-promoted alkyne-azide cycloaddition (SPAAC)