Learn More
Apexbio Technology LLC H-Leu-2-Chlorotrityl Resin 5g
SDP
Catalog No. 5000097354
Encompass_Preferred
H-Leu-2-Chlorotrityl Resin is a functionalized resin designed for solid-phase peptide synthesis (SPPS) It incorporates a leucine residue covalently linked via a chlorotrityl-based linker enabling controlled release of peptide intermediates with C-terminal leucine residues under acid-induced cleavage conditions Based on these properties H-Leu-2-Chlorotrityl Resin holds research potential in the synthesis and purification of leucine-terminated peptides supporting studies in pharmaceutically relevant peptide discovery bioactive peptide development and vaccine candidate research
Catalog No. 50-000-97354 Supplier Apexbio Technology LLC Supplier No. A68055000
H-Leu-2-Chlorotrityl Resin is a functionalized resin designed for solid-phase peptide synthesis (SPPS) It incorporates a leucine residue covalently linked via a chlorotrityl-based linker enabling controlled release of peptide intermediates with C-terminal leucine residues under acid-induced cleavage conditions Based on these properties H-Leu-2-Chlorotrityl Resin holds research potential in the synthesis and purification of leucine-terminated peptides supporting studies in pharmaceutically relevant peptide discovery bioactive peptide development and vaccine candidate research