Learn More
AdipoGen L-Leucine-7-amido-4-methylcoumarin hydrochloride
SDP
Catalog No. 501177566
Encompass_Preferred
L-Leucine-7-amido-4-methylcoumarin hydrochloride, AdipoGen Life Sciences. Chemical. CAS: 62480-44-8. Formula: C16H20N2O3 . HCl. MW: 324.8. Synthetic. Fluorogenic substrate for leucine aminopeptidase. Sensitivity of fluorescence detection is greatly increased over other methods such as 4-Methoxy-naphthylamide (pNA) assays. 62480-44-8
Catalog No. 50-117-7566 Supplier AdipoGen Supplier No. CDXL0003M010
L-Leucine-7-amido-4-methylcoumarin hydrochloride, AdipoGen Life Sciences. Chemical. CAS: 62480-44-8. Formula: C16H20N2O3 . HCl. MW: 324.8. Synthetic. Fluorogenic substrate for leucine aminopeptidase. Sensitivity of fluorescence detection is greatly increased over other methods such as 4-Methoxy-naphthylamide (pNA) assays. 62480-44-8