Learn More
Bachem H-Leu-Leu-Leu-OH
SDP
Supplier:  Bachem 4002312.0001
Encompass_Preferred
H-Leu-Leu-Leu-OH, 10329-75-6, C18H35N3O4, Mr 357.49, 1g. The tripeptide trileucine, found in casein and soy protein, was shown to increase the endothelial release of TXB2, a stable marker of TXA2. Inhibitor of human liver alanine aminopeptidase, Ki 0.054 mM. Addition of trileucine improved aerosol performance and stability of spray-dried powders for inhalation.
Catalog No. 50-259-786