Learn More
Bachem H-Leu-Leu-OMe · HBr
SDP
Supplier:  Bachem 4000725.0001
Encompass_Preferred
H-Leu-Leu-OMe · HBr, 16689-14-8, C13H26N2O3 · HBr, Mr 339.27, 1g. LLME · HBr. The lysosomotropic dipeptide LLME has selective toxicity for NK cells, CTL, and M phi without adverse effects on a variety of other lymphoid or nonlymphoid cell types.
Catalog No. NC9859913