Learn More
Bachem H-Lys(Abz)-Pro-Pro-pNA
SDP
Catalog No. 50260434
Encompass_Preferred
H-Lys(Abz)-Pro-Pro-pNA, 219138-18-8, C29H37N7O6, Mr 579.66, 50mg. Internally quenched fluorogenic (FRET) substrate which allows aminopeptidase P (APP) activity determination by a continuous fluorescent assay (kcat/Km = 29'000 M-1s-1 for E.coli APP and kcat/Km = 1'142'000 M-1s-1 for rat APP).
Catalog No. 50-260-434 Supplier Bachem Supplier No. 4027668.0050
H-Lys(Abz)-Pro-Pro-pNA, 219138-18-8, C29H37N7O6, Mr 579.66, 50mg. Internally quenched fluorogenic (FRET) substrate which allows aminopeptidase P (APP) activity determination by a continuous fluorescent assay (kcat/Km = 29'000 M-1s-1 for E.coli APP and kcat/Km = 1'142'000 M-1s-1 for rat APP).