Learn More
Apexbio Technology LLC H-Lys(Boc)-OH 2418-95-3 100g
SDP
Catalog No. 5000093525
Encompass_Preferred
H-Lys Boc -OH (2418-95-3) is a protected derivative of L-lysine utilized as a versatile synthetic intermediate It contains a tert-butyloxycarbonyl (Boc) group on the side-chain -amino functionality which temporarily masks this group to enable controlled and selective reactions during peptide synthesis and protein modification The Boc group can be removed under suitable conditions allowing the free amino group to participate in further conjugation or elongation steps This selective protection minimizes undesired cross-reactions enhancing specificity and purity in synthetic protocols Based on these properties H-Lys Boc -OH is extensively used in biochemical research therapeutic peptide development and protein structure function studies
Catalog No. 50-000-93525 Supplier Apexbio Technology LLC Supplier No. A7232100000
H-Lys Boc -OH (2418-95-3) is a protected derivative of L-lysine utilized as a versatile synthetic intermediate It contains a tert-butyloxycarbonyl (Boc) group on the side-chain -amino functionality which temporarily masks this group to enable controlled and selective reactions during peptide synthesis and protein modification The Boc group can be removed under suitable conditions allowing the free amino group to participate in further conjugation or elongation steps This selective protection minimizes undesired cross-reactions enhancing specificity and purity in synthetic protocols Based on these properties H-Lys Boc -OH is extensively used in biochemical research therapeutic peptide development and protein structure function studies