Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Lys(Fmoc)-OH | 84624-28-2 | Not available | 368.43 | 10 G
SDP

Catalog No. 5000448779
Encompass_Preferred

H-Lys(Fmoc)-OH is a lysine derivative intended for research use only. It is not sold for patient use. Amino acids and their derivatives, such as H-Lys(Fmoc)-OH, are widely used as ergogenic supplements, influencing the secretion of anabolic hormones, aiding fuel supply during exercise, enhancing mental performance under stress, and preventing exercise-induced muscle damage.

  • Molecular weight: 368.43
  • Formula: C21H24N2O4
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years; In solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-48779 Supplier Medchemexpress LLC Supplier No. HYW00819610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Lys(Fmoc)-OH is a lysine derivative intended for research use only. It is not sold for patient use. Amino acids and their derivatives, such as H-Lys(Fmoc)-OH, are widely used as ergogenic supplements, influencing the secretion of anabolic hormones, aiding fuel supply during exercise, enhancing mental performance under stress, and preventing exercise-induced muscle damage.

  • Molecular weight: 368.43
  • Formula: C21H24N2O4
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years; In solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.