Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Lys-Trp-Lys-OH | 38579-27-0 | 99.7% | 460.57 | 100 MG
SDP

Catalog No. 5000424771
Encompass_Preferred

H-Lys-Trp-Lys-OH is a small molecule peptide that shows antibacterial and antiviral activities. It was extracted from patent CN 104072579 A, Compound AMP12. This product is for research use only and not sold to patients.

  • Molecular weight: 460.57
  • Formula: C23H36N6O4
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: H-Lys-Trp-Lys-OH
  • Sequence shortening: HKWK-OH
  • Solubility in H2O: 100 mg/mL (217.12 mM; Needs ultrasonic)

Catalog No. 50-004-24771 Supplier Medchemexpress LLC Supplier No. HYP1350100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Lys-Trp-Lys-OH is a small molecule peptide that shows antibacterial and antiviral activities. It was extracted from patent CN 104072579 A, Compound AMP12. This product is for research use only and not sold to patients.

  • Molecular weight: 460.57
  • Formula: C23H36N6O4
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: H-Lys-Trp-Lys-OH
  • Sequence shortening: HKWK-OH
  • Solubility in H2O: 100 mg/mL (217.12 mM; Needs ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.