Learn More
Medchemexpress LLC H-Lys-Trp-Lys-OH | 38579-27-0 | 99.74% | 460.57 | 5 MG
SDP
Catalog No. 5000424699
Encompass_Preferred
H-Lys-Trp-Lys-OH is a small molecule peptide which displays antibacterial and antiviral activities, extracted from patent CN 104072579 A, Compound AMP12. It is for research use only and not sold to patients.
  • Small molecule peptide.
  • Displays antibacterial activity.
  • Displays antiviral activity.
  • For research use only.
Catalog No. 50-004-24699 Supplier Medchemexpress LLC Supplier No. HYP13505MG
H-Lys-Trp-Lys-OH is a small molecule peptide which displays antibacterial and antiviral activities, extracted from patent CN 104072579 A, Compound AMP12. It is for research use only and not sold to patients.
  • Small molecule peptide.
  • Displays antibacterial activity.
  • Displays antiviral activity.
  • For research use only.