Learn More
BIOHIPPO h/m GLUT1-sh-RFP/Luc
SDP
Catalog No. 5000331265
Encompass
h/m GLUT1-sh-RFP/Luc shRNA lentiviral vector expressing RFP/Luc with selection for studies in Human/Mouse Each vial provides a nominal titer of 2x10E6 transducing units TU Validated h/m GLUT1 shRNA Lentivirus High-titer shRNA lentiviral particles specific for human/mouse Glucose transporter 1 (GLUT1/GLUT-1/SLC2A1) The shRNA has been validated to meet or exceed 70% GLUT1 knockdown efficiency using a specific fluorescence-based method that is more rapid and reliable than qPCR The shRNA lentivirus is ultra-purified and concentrated to high-titer by PEG precipitation and sucrose gradient centrifugation and ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Order a shRNA lentivirus set and receive lentivirus produced from mix of 2 independent shRNAs validated to knockdown the target gene ( 70%) plus control lentivirus produced from mix of 2 scrambled shRNA constructs
Catalog No. 50-003-31265 Supplier BIOHIPPO Supplier No. BHV194001266
h/m GLUT1-sh-RFP/Luc shRNA lentiviral vector expressing RFP/Luc with selection for studies in Human/Mouse Each vial provides a nominal titer of 2x10E6 transducing units TU Validated h/m GLUT1 shRNA Lentivirus High-titer shRNA lentiviral particles specific for human/mouse Glucose transporter 1 (GLUT1/GLUT-1/SLC2A1) The shRNA has been validated to meet or exceed 70% GLUT1 knockdown efficiency using a specific fluorescence-based method that is more rapid and reliable than qPCR The shRNA lentivirus is ultra-purified and concentrated to high-titer by PEG precipitation and sucrose gradient centrifugation and ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Order a shRNA lentivirus set and receive lentivirus produced from mix of 2 independent shRNAs validated to knockdown the target gene ( 70%) plus control lentivirus produced from mix of 2 scrambled shRNA constructs