Learn More
BIOHIPPO h/m KLF5-sh-RFP-BSD
SDP
Catalog No. 5000331007
Encompass
h/m KLF5-sh-RFP-BSD shRNA lentiviral vector expressing RFP with Blasticidin selection for studies in Human/Mouse Each vial provides a nominal titer of 5x10E6 transducing units TU Validated h/m KLF5 shRNA Lentivirus High-titer shRNA lentiviral particles specific for human/mouse KLF transcription factor 5 (KLF5) The shRNA has been validated to meet or exceed 70% knockdown of KLF5 using a specific fluorescence-based method that is more rapid and reliable than qPCR The lentiviral particles are ultra-purified and concentrated by PEG precipitation and sucrose gradient centrifugation and are ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Order a shRNA lentivirus set and receive lentivirus produced from mix of 2 independent shRNAs validated to knockdown the target gene ( 70%) plus control lentivirus produced from mix of 2 scrambled shRNA constructs
Catalog No. 50-003-31007 Supplier BIOHIPPO Supplier No. BHV194001714
h/m KLF5-sh-RFP-BSD shRNA lentiviral vector expressing RFP with Blasticidin selection for studies in Human/Mouse Each vial provides a nominal titer of 5x10E6 transducing units TU Validated h/m KLF5 shRNA Lentivirus High-titer shRNA lentiviral particles specific for human/mouse KLF transcription factor 5 (KLF5) The shRNA has been validated to meet or exceed 70% knockdown of KLF5 using a specific fluorescence-based method that is more rapid and reliable than qPCR The lentiviral particles are ultra-purified and concentrated by PEG precipitation and sucrose gradient centrifugation and are ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Order a shRNA lentivirus set and receive lentivirus produced from mix of 2 independent shRNAs validated to knockdown the target gene ( 70%) plus control lentivirus produced from mix of 2 scrambled shRNA constructs