Learn More
BIOHIPPO h/m SREBP2-sh-RFP/Luc
SDP
Catalog No. 5000330864
Encompass
h/m SREBP2-sh-RFP/Luc shRNA lentiviral vector expressing RFP/Luc with selection for studies in Human/Mouse Each vial provides a nominal titer of 2x10E6 transducing units TU Validated h/m SREBP2 shRNA Lentivirus High-titer shRNA lentiviral particles specific for human/mouse sterol regulatory element binding transcription factor 2 (SREBP2/SREBF2) The shRNA has been validated to meet or exceed 70% knockdown of SREBP2 using a specific fluorescence-based method that is more rapid and reliable than qPCR The lentiviral particles are ultra-purified and concentrated by PEG precipitation and sucrose gradient centrifugation and are ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Order a shRNA lentivirus set and receive lentivirus produced from mix of 2 independent shRNAs validated to knockdown the target gene ( 70%) plus control lentivirus produced from mix of 2 scrambled shRNA constructs
Catalog No. 50-003-30864 Supplier BIOHIPPO Supplier No. BHV194001706
h/m SREBP2-sh-RFP/Luc shRNA lentiviral vector expressing RFP/Luc with selection for studies in Human/Mouse Each vial provides a nominal titer of 2x10E6 transducing units TU Validated h/m SREBP2 shRNA Lentivirus High-titer shRNA lentiviral particles specific for human/mouse sterol regulatory element binding transcription factor 2 (SREBP2/SREBF2) The shRNA has been validated to meet or exceed 70% knockdown of SREBP2 using a specific fluorescence-based method that is more rapid and reliable than qPCR The lentiviral particles are ultra-purified and concentrated by PEG precipitation and sucrose gradient centrifugation and are ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Order a shRNA lentivirus set and receive lentivirus produced from mix of 2 independent shRNAs validated to knockdown the target gene ( 70%) plus control lentivirus produced from mix of 2 scrambled shRNA constructs