Learn More
BIOHIPPO h/m USP20 shRNA Set GFP Puro
SDP
Catalog No. 5000331415
Encompass
h/m USP20 shRNA Set (GFP Puro) shRNA lentiviral vector expressing GFP with Puromycin selection for studies in Human/Mouse Each vial provides a nominal titer of 5x10E6 (sh-mix) 5x10E6 (scr-mix) transducing units TU Validated human/mouse USP20 shRNA Lentivirus High-titer shRNA lentiviral particles specific for human/mouse ubiquitin specific peptidase 20 (USP20) The shRNA has been validated to meet or exceed 70% USP20 knockdown efficiency using a specific fluorescence-based method that is more rapid and reliable than qPCR The shRNA lentivirus is ultra-purified and concentrated to high-titer by PEG precipitation and sucrose gradient centrifugation and ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Order a shRNA lentivirus set and receive lentivirus produced from mix of 2 independent shRNAs validated to knockdown the target gene ( 70%) plus control lentivirus produced from mix of 2 scrambled shRNA constructs
Catalog No. 50-003-31415 Supplier BIOHIPPO Supplier No. BHV194001461
h/m USP20 shRNA Set (GFP Puro) shRNA lentiviral vector expressing GFP with Puromycin selection for studies in Human/Mouse Each vial provides a nominal titer of 5x10E6 (sh-mix) 5x10E6 (scr-mix) transducing units TU Validated human/mouse USP20 shRNA Lentivirus High-titer shRNA lentiviral particles specific for human/mouse ubiquitin specific peptidase 20 (USP20) The shRNA has been validated to meet or exceed 70% USP20 knockdown efficiency using a specific fluorescence-based method that is more rapid and reliable than qPCR The shRNA lentivirus is ultra-purified and concentrated to high-titer by PEG precipitation and sucrose gradient centrifugation and ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Order a shRNA lentivirus set and receive lentivirus produced from mix of 2 independent shRNAs validated to knockdown the target gene ( 70%) plus control lentivirus produced from mix of 2 scrambled shRNA constructs