Learn More
Bioworld H Medium Broth, 500 g
Catalog No. 50254469
Encompass_Preferred
H Medium BrothBiotechnology Grade pH: 6.8-7.2 Dehydrated Appearance: light beige powder Soluble in waterMedia Formulation (g/L): 10g Tryptone, 8g Sodium chloride (pH @ 25°C 7 ±0.2)
Catalog No. 50-254-469 Supplier Bioworld Supplier No. 306260003
H Medium BrothBiotechnology Grade pH: 6.8-7.2 Dehydrated Appearance: light beige powder Soluble in waterMedia Formulation (g/L): 10g Tryptone, 8g Sodium chloride (pH @ 25°C 7 ±0.2)