Learn More
Bachem H-Met-Met-OH
SDP
Supplier:  Bachem 4000202.0001
Encompass_Preferred
H-Met-Met-OH, 7349-78-2, C10H20N2O3S2, Mr 280.41, 1g. Yang et al. observed that methionyl-methionine promotes a-s1 casein synthesis in bovine mammary gland explants.
Catalog No. 50-259-364