Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Met-Met-OH (L-Methionyl-L-methionine) | 7349-78-2 | 98.11% | 280.41 | 1 ML
SDP

Catalog No. 5000361721
Encompass_Preferred

H-Met-Met-OH (L-Methionyl-L-methionine) is a dipeptide composed of two methionine residues. It functions as a dietary supplement that has demonstrated various health benefits in animals and is orally active.

  • Composed of two methionine residues
  • Dietary supplement
  • Orally active
  • Appearance: solid, colorless to off-white
  • Soluble in DMSO at 100 mg/mL (ultrasonic required)
  • Achieves ≥ 2.5 mg/mL solubility in various in vivo solvent systems

Catalog No. 50-003-61721 Supplier Medchemexpress LLC Supplier No. HY15001310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Met-Met-OH (L-Methionyl-L-methionine) is a dipeptide composed of two methionine residues. It functions as a dietary supplement that has demonstrated various health benefits in animals and is orally active.

  • Composed of two methionine residues
  • Dietary supplement
  • Orally active
  • Appearance: solid, colorless to off-white
  • Soluble in DMSO at 100 mg/mL (ultrasonic required)
  • Achieves ≥ 2.5 mg/mL solubility in various in vivo solvent systems

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.