Learn More
Medchemexpress LLC H-Met-oipr hydrochloride | 85391-05-5 | MFCD00190954 | 97.0% | 227.75 g/mol | C8H18ClNO2S | 25 G
SDP
Catalog No. 5000303492
Encompass_Preferred
H-Met-OiPr hydrochloride (CAS 85391-05-5) is the isopropyl ester hydrochloride derivative of methionine supplied for research use. It is used as an amino acid building block in synthetic chemistry, including preparation of farnesyl-protein transferase inhibitor intermediates.
  • High purity, typically 97.0%.
  • Molecular formula C8H18ClNO2S and molecular weight 227.75 g/mol.
  • Useful as an amino acid derivative for synthesis and research applications.
  • Provided as a solid reagent; commonly available in 25 g packs.
  • For research use only; not for human or clinical use.
Catalog No. 50-003-03492 Supplier Medchemexpress LLC Supplier No. 25GHYW022657
H-Met-OiPr hydrochloride (CAS 85391-05-5) is the isopropyl ester hydrochloride derivative of methionine supplied for research use. It is used as an amino acid building block in synthetic chemistry, including preparation of farnesyl-protein transferase inhibitor intermediates.
  • High purity, typically 97.0%.
  • Molecular formula C8H18ClNO2S and molecular weight 227.75 g/mol.
  • Useful as an amino acid derivative for synthesis and research applications.
  • Provided as a solid reagent; commonly available in 25 g packs.
  • For research use only; not for human or clinical use.