Learn More
Medchemexpress LLC H-Met-OtBu.HCl | 91183-71-0 | 99.7% | 241.78 | 5 G
SDP
Catalog No. 5000366910
Encompass_Preferred
H-Met-OtBu.HCl is a Methionine derivative for research use only. It is utilized as an ergogenic supplement, influencing the secretion of anabolic hormones, the supply of fuel during exercise, and mental performance in stressful tasks. It also helps prevent exercise-induced muscle damage.
  • Impacts the secretion of anabolic hormones.
  • Supports fuel supply during exercise.
  • Enhances mental performance during stressful tasks.
  • Prevents exercise-induced muscle damage.
Catalog No. 50-003-66910 Supplier Medchemexpress LLC Supplier No. HYW0095625G
H-Met-OtBu.HCl is a Methionine derivative for research use only. It is utilized as an ergogenic supplement, influencing the secretion of anabolic hormones, the supply of fuel during exercise, and mental performance in stressful tasks. It also helps prevent exercise-induced muscle damage.
  • Impacts the secretion of anabolic hormones.
  • Supports fuel supply during exercise.
  • Enhances mental performance during stressful tasks.
  • Prevents exercise-induced muscle damage.