Learn More
Fine Science Tools Halsted-Mosquito Hemostats (Left-Handed), Stainless Steel, Curved, 12.5 cm
SDP
Catalog No. NC1741526
Encompass
Tip Shape: Curved/ Tips: Serrated/ Tip Width: 1mm/ Clamping Length: 20mm/ Alloy / Material: Stainless Steel/ Length: 12.5cm
Catalog No. NC1741526 Supplier Fine Science Tools Supplier No. 1310912
Tip Shape: Curved/ Tips: Serrated/ Tip Width: 1mm/ Clamping Length: 20mm/ Alloy / Material: Stainless Steel/ Length: 12.5cm