Learn More
Apexbio Technology LLC H-N-Me-Ser-OH.HCl 2480-26-4 5g
SDP
Catalog No. 5000094147
Encompass_Preferred
H-N-Me-Ser-OH HCl (CAS 2480-26-4) is a methyl-substituted derivative of serine used as a synthetic building block in peptide chemistry It is employed to investigate the molecular consequences of serine methylation in biologically active peptides and proteins This compound serves as a substrate analog or probe in biochemical and structural research facilitating elucidation of the functional impact of site-specific amino acid modifications including protein methylation enzyme catalysis mechanisms and protein interaction pathways Based on these properties H-N-Me-Ser-OH HCl has potential applications in studies of protein modification enzyme function and protein protein interaction analysis
Catalog No. 50-000-94147 Supplier Apexbio Technology LLC Supplier No. A77135000
H-N-Me-Ser-OH HCl (CAS 2480-26-4) is a methyl-substituted derivative of serine used as a synthetic building block in peptide chemistry It is employed to investigate the molecular consequences of serine methylation in biologically active peptides and proteins This compound serves as a substrate analog or probe in biochemical and structural research facilitating elucidation of the functional impact of site-specific amino acid modifications including protein methylation enzyme catalysis mechanisms and protein interaction pathways Based on these properties H-N-Me-Ser-OH HCl has potential applications in studies of protein modification enzyme function and protein protein interaction analysis