Learn More
Bachem H-Nle-Arg-Phe-NH2
SDP
Supplier:  Bachem 4009465.0100
Encompass_Preferred
H-Nle-Arg-Phe-NH2 Acetate, 104809-29-2, C21H35N7O3, Mr 433.55, 100mg. nLRFamide, synthetic analog of flatworm FMRFamide-related peptides (FaRPs) showed considerable myoactivity on isolated muscle fibers of the human blood fluke.
Catalog No. 50-259-676